Learn More
Medchemexpress LLC HY-14121 5mg Medchemexpress, MRS 1754 CAS:264622-58-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY141215MG
Encompass_Preferred
Medchemexpress, HY-14121 5mg MRS 1754 CAS:264622-58-4 MRS 1754 is a selective antagonist radioligand for A 2B adenosine receptor with very low affinity for A 1 and A 3 receptors of both humans and rats [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2694