Learn More
Medchemexpress LLC MRT-83 hydrochloride | 1359944-60-7 | 99.6% | 575.05 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1828710MM/1ML
Encompass_Preferred
MRT-83 hydrochloride is a potent antagonist of the Smoothened (Smo) receptor, known for inhibiting the Hedgehog (Hh) signaling pathway. It shows promise for researching various cancer diseases, demonstrating specific inhibitory concentrations in cell assays.
  • Potent antagonist of Smoothened (Smo) receptor
  • Inhibits Hedgehog (Hh) signaling pathway
  • Inhibits BODIPY-cyclopamine binding to human Smo
  • Potential for researching cancer diseases
  • Exhibits antiproliferative activity against human HT1080 and LS180 cells
Catalog No. 50-003-87554