Learn More
Cayman Chemical MRT67307hydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1991610 MG
Encompass_Preferred
A kinase inhibitor that inhibits TBK1, MARK1-4, IKKε, and NUAK1 (IC50 values are 19, 27-52, 160, and 230 nM, respectively), SIKs (IC50s = 250, 67, and 430 nM for SIK1, SIK2, and SIK3, respectively) and ULK1 and ULK2 (IC50s = 45 and 38 nM, respectively)
Catalog No. 50-298-1877