Learn More
Medchemexpress LLC MRT68921 dihydrochloride | 2080306-21-2 | 99.7% | 507.50 | C25H36Cl2N6O | 1 ML
SDP
Supplier:  Medchemexpress LLC HY100006A10MM 1ML
Encompass_Preferred
MRT68921 dihydrochloride is a potent dual inhibitor of ULK1/ULK2 and NUAK1 used in research to block autophagy and probe kinase signaling in cellular and biochemical assays. Supplied as a DMSO solution (10 mM, 1 mL) and as solid quantities, the product has high purity and defined storage conditions for reproducible experimental use.
  • Potent dual ULK1/ULK2 and NUAK1 inhibition for autophagy studies.
  • Available as a 10 mM solution in DMSO and multiple solid sizes for flexible dosing.
  • High purity (≈99.7%) suitable for biochemical and cellular assays.
  • Characterized by molecular formula and molecular weight for identity confirmation.
  • Defined storage recommendations to preserve stability in solvent and solid form.
Catalog No. 50-002-15104