Learn More
Medchemexpress LLC MRT68921 dihydrochloride | 2080306-21-2 | 99.7% | 507.50 | C25H36Cl2N6O | 50 MG

Supplier: Medchemexpress LLC HY100006A50MG
MRT68921 dihydrochloride is a potent small-molecule inhibitor of ULK1/ULK2 and NUAK1 used in biochemical and cellular research to probe autophagy and kinase signaling. Supplied as a high-purity solid for laboratory research applications only.
- Potent NUAK1/ULK1 dual inhibitor.
- Suitable for biochemical and cell-based assays.
- High purity: 99.7%.
- Molecular weight: 507.50.
- Storage: solid at 4°C; solutions -80°C for long-term.
- Appearance: white to off-white solid.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.