Learn More
Cayman Chemical MRtnsIn PolycLNal ntIbod 100ug
SDP
Supplier:  Cayman Chemical 41262100 UG
Encompass_Preferred
Immunogen: Mertansine-KLH conjugate • Host: Rabbit • Species Reactivity: Species-independent • Cross Reactivity: (+) Mertansine-conjugated antibodies; (-) Albumin, VcMMAE-conjugated antibodies, unconjugated antibodies • Applications: ELISA, WB
Catalog No. 50-294-4495