Learn More
Apexbio Technology LLC MRTX-1133(Synonyms: MRTX1133, MRTX 1133, Mirati-1133, KRAS G12D inhibitor MRTX1133), 1mg, CAS: 2621928-55-8.
SDP
Supplier:  Apexbio Technology LLC B87611
Encompass_Preferred
MRTX-1133 (CAS 2621928-55-8) is a non-covalent alkyne-based inhibitor specifically targeting the KRAS G12D mutant protein It binds KRAS G12D with sub-picomolar affinity occupying the Switch II pocket and forming multiple intermolecular interactions Through these interactions MRTX-1133 disrupts SOS1-mediated guanine nucleotide exchange and prevents the assembly of KRAS-GTP/RAF1 signaling complexes thereby inhibiting downstream KRAS-driven signaling pathways MRTX-1133 is applied in cancer biology research to investigate tumor signaling and therapeutic strategies involving KRAS G12D mutations
Catalog No. 50-000-61040