Learn More
Medchemexpress LLC MRTX849 acid | 2561529-96-0 | MFCD00868113 | 99.8% | 662.15 | C34H37ClFN7O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1394025MG
Encompass_Preferred
MRTX849 acid is the carboxylic acid derivative of MRTX849 used as a ligand or synthetic intermediate in KRAS G12C research and PROTAC preparation. It is supplied as a high-purity, light yellow solid with molecular formula C34H37ClFN7O4 and molecular weight 662.15. Store at -20°C and protect from light; in solution, store at -80°C for long-term or -20°C for short-term.
  • High purity suitable for research applications.
  • Light yellow solid facilitating handling.
  • Useful as a ligand or intermediate for KRAS G12C and PROTAC synthesis.
  • Stable when stored at -20°C and protected from light.
  • Storage recommendations provided for neat material and solutions.
Catalog No. 50-002-22689