Learn More
Medchemexpress LLC MRX-2843 | 1429882-07-4 | MFCD28502224 | 99.6% | 488.67 g/mol | C29H40N6O | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10154950MG
Encompass_Preferred
MRX-2843 (UNC2371) is an orally active, ATP-competitive dual MERTK and FLT3 tyrosine kinase inhibitor used in preclinical cancer research. Reported enzymatic IC50 values are 1.3 nM for MERTK and 0.64 nM for FLT3. Supplied as a solid with high chemical purity, it is intended for in vitro and in vivo studies of kinase signaling and mechanisms of drug resistance.
  • Potent dual inhibition of MERTK and FLT3 with low-nanomolar potency.
  • Orally bioavailable type 1 kinase inhibitor suitable for preclinical models.
  • High chemical purity supports reproducible research results.
  • Supplied as a solid for flexible formulation and dosing.
  • Useful for studying kinase signaling pathways and resistance mechanisms.
Catalog No. 50-002-48951