Learn More
Medchemexpress LLC MRX-2843 (UNC2371) | 1429882-07-4 | MFCD28502224 | 99.6% | 488.67 g/mol | C29H40N6O | 25 MG

Supplier: Medchemexpress LLC HY10154925MG
MRX-2843 (UNC2371) is an orally active, ATP-competitive small-molecule inhibitor of the MERTK and FLT3 receptor tyrosine kinases used in preclinical research to study signaling, overcome resistance-conferring FLT3 mutations, and evaluate anti-leukemia effects. Analytical data and handling recommendations are provided for laboratory use.
- Potent dual MERTK and FLT3 enzymatic inhibition (IC50 1.3 nM and 0.64 nM).
- Demonstrated activity in cell proliferation assays (Kasumi-1 IC50 ≈ 143.5 nM).
- High purity suitable for research use (99.6% by HPLC).
- Orally active small molecule compatible with in vivo studies.
- Supplied as solid and solution formulations for flexible handling.
- Recommended storage: powder at -20°C or 4°C; in solvent at -80°C or -20°C.
- Molecular weight 488.67 g/mol; chemical formula C29H40N6O.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.