Learn More
Medchemexpress LLC Ms-444 | 150045-18-4 | MFCD00923100 | 99.5% | 230.22 g·mol⁻¹ | C13H10O4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100685100MG
Encompass_Preferred
MS-444 is a small-molecule inhibitor of myosin light chain kinase (MLCK) and the RNA-binding protein HuR (ELAVL1). It inhibits purified smooth muscle MLCK with an IC50 of approximately 10 μM and produces growth inhibition in multiple colorectal cancer cell lines; it is used in biochemical assays, cell-based studies, and preclinical in vivo research.
Catalog No. 50-002-12632
Add to Cart
Request a Quote