Learn More
Medchemexpress LLC Ms-444 | 150045-18-4 | MFCD00923100 | 99.5% | 230.22 g/mol | C13H10O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10068525MG
Encompass_Preferred
MS-444 is a small-molecule research inhibitor that targets smooth muscle myosin light chain kinase (MLCK) and the RNA-binding protein HuR (ELAVL1). It inhibits MLCK with an IC50 of 10 μM and shows growth inhibition in colorectal cancer cell lines with IC50 values of 5.60-14.21 μM. The compound is supplied as a high-purity solid and is intended for laboratory research use only.
  • Inhibits MLCK with IC50 of 10 μM
  • Inhibits HuR and reduces growth in colorectal cancer cell lines (IC50 5.60-14.21 μM)
  • High purity suitable for research applications
  • Offered as a solid with small-scale quantities for lab experiments
  • Store solid at -20°C, protect from light, under inert gas
  • Molecular weight 230.22 g/mol; formula C13H10O4
Catalog No. 50-002-13692