Learn More
Biocare Medical, LLC Mouse-on-Mouse HRP-Polymer Bundle - 100 ml
SDP
Supplier:  Biocare Medical, LLC MM510L
Encompass_Preferred
The Mouse-on-Mouse HRP-Polymer is specially designed for using mouse primary antibodies on mouse tissues with minimum cross-reactivity to endogenous mouse IgG. Utilization of this goat anti-mouse horseradish peroxidase (HRP) polymer is advantageous over traditional biotinylation methods. Biotinylated anti-mouse secondary antibodies can bind to endogenous mouse IgG in the tissue in addition to the primary antibody, leading to troublesome background staining. Biocare Medical's biotin-free polymer technology provides the elimination of endogenous biotin, reduction of IHC steps and increased sensitivity.
Catalog No. 50-829-33