Learn More
Innovative Research Inc Ms PAI1 Rcmb WT Act 500ug
Supplier:  Innovative Research Inc IMSPAI1ARWT500UG
Encompass_Preferred
Mouse PAI-1 Recombinant Wild Type Active 500 ug - Mouse PAI-1 Recombinant Wild Type Active from Innovative Research has been recombinantly produced in E. coli and purified by affinity chromatography. This is a Frozen liquid buffered in 0.05M Sodium Acetate; 0.4M NaCl; 1mM EDTA; pH 5.0 and a purity of >95% as determined by SDS-PAGE. More Details Species Mouse Target PAI-1 Purification Method Affinity purified Purity 95% as determined by SDS-PAGE Source E. coli Storage Conditions -70 C Additional Information Active fraction contains between 30% and 70% active PAI-1. This product can be used as a control in applications like ELISA and Western Blot. It can be used for a wide range of in vitro applications, including fibrinolysis, hypertension studies, inhibition, plasminogen activation, protein-protein interactions, receptor binding, and more. For Research Use Only.
Catalog No. 50-274-4144