Learn More
Innovative Research Inc Ms uPA Act Rcmb HMW 50ug
Supplier:  Innovative Research Inc IMSUPAARCHMW50UG
Encompass_Preferred
Mouse Urokinase (uPA) Active Recombinant High Molecular Weight 50ug - Mouse Urokinase (uPA) Active Recombinant High Molecular Weight from Innovative Research has been recombinantly produced in insect cells and purified using a combination of conventional and affinity chromatography. This is a frozen liquid buffered in 0.05M Sodium Acetate; 0.1M NaCl; 1mM EDTA; pH 5.0 and a purity of >95%. This product is useful as a positive control for ELISA and western blot and for a variety of in vitro applications including cell differentiation, cell signaling, clotting assays, fibrinolysis, plasminogen activation, protein-protein interactions, receptor binding, thromboelastography, and thrombolysis. Active two-chain high molecular weight mouse urokinase. More Details Source Insect Cell Culture Purification Chromatographically purified Purity >95% Storage Conditions -70 C
Catalog No. 50-274-4130