Learn More
Innovative Research Inc Ms uPA Act Rcmb LMW 50ug
Supplier:  Innovative Research Inc IMSUPAARCLMW50UG
Encompass_Preferred
Mouse Urokinase (uPA) Active Recombinant Low Molecular Weight 50ug - There is currently an extended lead time for this product. Orders placed today will be filled when material becomes available. Mouse Urokinase (uPA) Active Recombinant Low Molecular Weight from Innovative Research has been recombinantly produced in insect cells and purified using a combination of conventional and affinity chromatography. This is a frozen liquid buffered in 0.05M Sodium Acetate; 0.1M NaCl; 1mM EDTA; pH 5.0 and a purity of >95%. This product is useful as a positive control for ELISA and western blot and for a variety of in vitro applications including cell differentiation, cell signaling, clotting assays, fibrinolysis, plasminogen activation, protein-protein interactions, receptor binding, thromboelastography, and thrombolysis. Active two-chain low molecular weight mouse urokinase. More Details Source Insect Cell Culture Purification Chromatographically purified Purity >95% Storage Conditions -70
Catalog No. 50-274-4129