Learn More
Waters Corp Waters Corporation MS UPLC Columns Method Development Kit, 2.1 mm, 100 mm, 4 per pack
Supplier: Waters Corp 176001608
This MS UPLC method development kit combines diverse column chemistries (1.7 μm BEH C18, 1.8 μm HSS C18, C18 SB, and T3) for efficient and effective method development. Leveraging UPLC Technology, it improves throughput and reduces analysis time, accelerating product market entry and improving assay profitability.
- Kit contains four columns offering varied selectivities.
- Features distinct silanol activity, shape, selectivity, and hydrophobicity; no MS bleed.
- Reversed Phase separation mode, pH range 1 to 12.
- Maximum pressure: 18000 psi (1240 Bar).
- Spherical particle shape, 100 Å pore size.
- Compatible with UPLC and UHPLC systems.
- Dimensions: 2.1 mm inner diameter, 100 mm length.
- Endcapped, low silanol activity, Parker-style endfitting, eCord technology.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.