Learn More
TARGETMOL CHEMICALS INC MS023 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T690010MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MS023 is a potent selective and cell-active Type I PRMT inhibitor with IC50 of 30 nM 119 nM 83 nM 4 nM and 5 nM for PRMT1 PRMT3 PRMT4 PRMT6 and PRMT8 respectively. purity: 99%
Catalog No. 50-257-6419