Learn More
Medchemexpress LLC MS023 | 1831110-54-3 | 99.9% | 287.40 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY19615100MG
Encompass_Preferred
MS023 is a chemical probe and a potent, selective, and cell-active inhibitor of human type I protein arginine methyltransferases (PRMTs), with IC50s of 30, 119, 83, 4, and 5 nM for PRMT1, PRMT3, PRMT4, PRMT6, and PRMT8. It is utilized for research in epigenetics, particularly in studies involving histone methyltransferases, and demonstrates antiproliferative effects against human A549, HCC4006, and SK-LU-1 cells.
  • Potent, selective, and cell-active inhibitor of human type I protein arginine methyltransferases (PRMTs)
  • Inhibits PRMT1 methyltransferase activity in MCF7 cells
  • Inhibits PRMT6 methyltransferase activity in HEK293 cells
  • Blocks MLL-r acute lymphoblastic leukemia (ALL) propagation in animal models
  • Extends survival of leukemic mice in combination treatments
Catalog No. 50-004-01282