Learn More
Selleck Chemical LLC MS023 5mg 1831110-54-3
SDP
Supplier:  Selleck Chemical LLC S81125MG
Encompass_Preferred
MS023 is a potent, selective, and cell-active Type I PRMT inhibitor with IC50 of 30 nM, 119 nM, 83 nM, 4 nM, and 5 nM for PRMT1, PRMT3, PRMT4, PRMT6 and PRMT8, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5666