Learn More
Selleck Chemical LLC MS023 hydrochloride E2887-5MG
SDP
Supplier:  Selleck Chemical LLC E28875MG
Encompass_Preferred
MS023 hydrochloride(Compound 3) is a potent selective and cell-active inhibitor of human type I protein arginine methyltransferases (PRMT) with IC50 values of 30 119 83 4 and 5 nM for PRMT1 PRMT3 PRMT4 PRMT6 and PRMT8 respectively
Catalog No. 50-000-40611