Learn More
ATCC MS1; Pancreas; Mouse (Mus musculus)
Supplier:  ATCC CRL2279
Encompass
MS1 (MILE SVEN 1) is an endothelial cell that was isolated from the islet of Langerhans of a mouse. This cell line can be used for the study of signal transduction of angiogenic factors. The cell was deposited by JL Arbiser in 1994.
Catalog No. 50-238-1962
 Restricted Item