Learn More
Medchemexpress LLC MS31 (trihydrochloride) | 2748239-18-9 | 98.0% | 450.83 g/mol | C20H30Cl3N3O2 | 100MG

Supplier: Medchemexpress LLC HY125837A100MG
MS31 trihydrochloride is a fragment-like small-molecule inhibitor of the methyllysine reader protein Spindlin 1 (SPIN1), supplied as the trihydrochloride salt for research use. The compound exhibits low-nanomolar affinity for SPIN1 and is provided as a white to off-white solid with high purity suitable for biochemical and binding assays.
- High purity (98.0%) suitable for biochemical studies.
- Molecular weight 450.83 g/mol and formula C20H30Cl3N3O2.
- Demonstrated SPIN1 activity: AlphaLISA IC50 77 nM; FP IC50 243 nM; Kd 91 nM.
- Soluble in DMSO at 62.5 mg/mL (138.63 mM); ultrasonic assistance recommended.
- Stable as powder at 4°C; solutions stable at -80°C (6 months) or -20°C (1 month) when sealed.
- Available in multiple pack sizes, including 100 mg, for assay development and screening.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.