Learn More
Medchemexpress LLC MS4078 50mg | 2229036-62-6 | 99.7% | 914.47 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11215550MG
Encompass_Preferred
MS4078 is an anaplastic lymphoma kinase (ALK) PROTAC (degrader) based on Cereblon ligand, demonstrating a binding affinity to ALK with a Kd of 19 nM. This compound is intended for research use only. It has been shown to effectively inhibit cancer cell proliferation and potently reduce ALK fusion protein levels.
  • Effectively inhibits cancer cell proliferation.
  • Potently reduces ALK fusion protein levels.
  • Inhibits ALK auto-phosphorylation.
  • Inhibits downstream STAT3 phosphorylation.
Catalog No. 50-004-35700