Learn More
Medchemexpress LLC HY-130602 5mg , MS432 CAS:2672512-44-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1306025MG
Encompass_Preferred
HY-130602 5mg MS432 CAS: 2672512-44-4 MS432 is a first-in-class and highly selective PD0325901-based von Hippel-Lindau-recruiting PROTAC degrader for MEK1 and MEK2. MS432 displays good plasma exposure in mice, exhibiting DC50 values of 31 nM and 17 nM for MEK1, MEK2 in HT29 cells respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0034