Learn More
Medchemexpress LLC MS4322 (isomer) | 2601727-80-2 | 99.9% | C55H76N10O12S | 1 MG
SDP
Supplier:  Medchemexpress LLC HY141877B1MG
Encompass_Preferred
MS4322 (YS43-22) isomer is an isomer of MS4322. It is a specific PRMT5 PROTAC degrader. It can be used for the research of breast cancer, lung cancer, and hepatocellular cancer.
  • Reduces the PRMT5 protein level with a DC50 of 1.1 μM in MCF-7 cells.
  • Inhibits the methyltransferase activity of PRMT5 with an IC50 of 18 nM.
  • Promotes ubiquitination and degradation of PRMT5.
Catalog No. 50-003-53630