Learn More
Medchemexpress LLC MS437 5mg | 541535-31-3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1149945MG
Encompass_Preferred
MS437 is a research-grade small molecule that potently stimulates the thyroid-stimulating hormone receptor (TSHR), activating Gsα, Gαq, and Gα12 signaling and up-regulating thyroglobulin, the sodium iodide symporter, and TSHR gene expression. It is supplied as a high-purity solid for biochemical and cell-based research applications.
  • High purity (98.0%).
  • Molecular formula C22H24N2O; molecular weight 332.44 g·mol⁻1.
  • Potent TSH receptor stimulation (EC50 ≈ 1.3 x 10⁻7 M).
  • Available in multiple milligram sizes, including 5 MG.
  • Suitable for biochemical assays and cell-based receptor pharmacology studies.
Catalog No. 50-002-27723