Learn More
Medchemexpress LLC MS48107 | 2375070-79-2 | MFCD03820803 | 99.9% | 417.44 | C23H20FN5O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1344945MG
Encompass_Preferred
MS48107 is a research small molecule characterized as a potent and selective positive allosteric modulator of the proton-sensing G protein-coupled receptor GPR68. It is supplied for laboratory research use to probe GPR68 signaling in biochemical, cellular, and some in vivo studies; datasheet and COA provide physicochemical data and handling recommendations.
  • Potent and selective positive allosteric modulator of GPR68.
  • Suitable for in vitro and in vivo research to study GPCR signaling.
  • High purity: 99.9% (LCMS) for supplied batches.
  • Molecular formula C23H20FN5O2; molecular weight 417.44.
  • Stable as powder at -20°C for long-term storage; follow recommended solvent storage conditions.
Catalog No. 50-002-22947