Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MS7972 | 2165322-94-9 | 98.0% | 422.21 | C17H16BrF4NO2 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY11737050MG

Encompass_Preferred

USP25/28 inhibitor AZ1 is a selective, orally active, noncompetitive small-molecule inhibitor of deubiquitinases USP25 and USP28, used for cellular and in vivo studies to probe deubiquitination pathways and disease models.

  • Selective dual USP25 and USP28 inhibitor.
  • Submicromolar potency: IC50 0.7 μM (USP25), 0.6 μM (USP28).
  • High purity: 98.0%.
  • Solid powder form suitable for in vitro and in vivo applications.
  • Storage recommendations: powder -20°C long term, 4°C short term; in solvent -80°C for extended storage.

Catalog No. 50-002-26928


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.