Learn More
Medchemexpress LLC MS8511 hydrochloride | 3031788-28-7 | C28H42ClN5O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY150510A5MG
Encompass_Preferred
MS8511 hydrochloride is an off-white to light yellow solid. It is a specific, covalent, and irreversible inhibitor of G9a/GLP, targeting the cysteine residue at the substrate binding site.
  • Specific covalent and irreversible inhibitor of G9a/GLP
  • Targets the cysteine residue at the substrate binding site
  • IC50 of 100 nM for G9a and 140 nM for GLP
  • Kd values of 44 nM for G9a and 46 nM for GLP
  • Reduces intracellular H3K9me2 levels
  • Enhances antiproliferative activity
  • Useful for studying various cancers and other diseases like Alzheimer's disease, sickle cell disease, and Prader-Willi syndrome
Catalog No. 50-003-75273