Learn More
Apexbio Technology LLC MSAB 173436-66-3 100mg
SDP
Supplier:  Apexbio Technology LLC A8832100
Encompass_Preferred
MSAB (CAS 173436-66-3) is a selective small molecule inhibitor that targets the Wnt/ -catenin signaling pathway It acts by directly binding to the -catenin protein thereby promoting its degradation and suppressing the downstream transcription of Wnt pathway target genes Studies have shown that MSAB reduces the transcriptional activity associated with Wnt/ -catenin signaling and inhibits the growth of tumor cells reliant on this pathway MSAB is widely utilized in research to investigate the molecular mechanisms of Wnt-dependent cancers and to facilitate the development of anti-tumor therapeutics targeting this signaling cascade
Catalog No. 50-000-96010