Learn More
Medchemexpress LLC MSC-1186 | 2871698-23-4 | 98.95% | 461.90 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14851810MM/1ML
Encompass_Preferred
MSC-1186 is a chemical probe and a highly selective pan-SRPK inhibitor. It demonstrates activity for SRPK1, SRPK2, and SRPK3 with IC50 values of 2.7 nM, 81 nM, and 0.6 nM, respectively. This compound can be utilized in cancer research.
  • Highly selective pan-SRPK inhibitor
  • Active against SRPK1, SRPK2, and SRPK3
  • Can be used for cancer research
  • Exhibits excellent kinome-wide selectivity
  • Shows additive attenuation of SR-protein phosphorylation when used with CLK inhibitors
Catalog No. 50-003-61149