Learn More
Meso Scale Discovery MSD GOLD 96-well Small Spot Streptavidin SECTOR Plate, 1 Plate
SDP
Supplier:  Meso Scale Discovery L45SA1
Encompass
MSD GOLD Small Spot Streptavidin plates bind biotin-conjugated molecules for flexible and rapid assay development. MSD GOLD designation certifies that the plates and reagents conform to rigorous specifications for uniformity and precision. These specifications are designed to meet the requirements of critical assays and clinical research for the drug safety and toxicology communities. Small Spot Streptavidin Plates provide high assay signals and therefore superior sensitivity. These plates exhibit lower non-specific binding, especially with complex sample matrices. They may be used for a range of applications including immunogenicity testing, pharmacokinetic / pharmacodynamic studies, and biomarker assay development, and screening.
Catalog No. NC0889933