Learn More
Vector Laboratories Monoclonal Mouse anti‑Human ANO1 / DOG1 / TMEM16A Antibody (clone DOG‑1.1, aa904‑986, IHC) LS‑C391534
SDP
Supplier:  Vector Laboratories LSC391534100
Encompass
TMEM16A antibody LS-C391534 is an unconjugated mouse monoclonal antibody to human TMEM16A (ANO1 / DOG1) (aa904-986). Validated for IHC.
Catalog No. NC3715583