Learn More
Raybiotech Inc MSE INFLAMMATION ARRAY C1 (8)
Supplier:  Raybiotech Inc AAMINF18
Encompass_Preferred
RayBio C-Series Mouse Inflammation Antibody Array 1 Kit. Detects 40 Mouse Inflammatory Factors. Suitable for all liquid sample types.
Catalog No. 50-104-3236