Learn More
Lonza Walkersville MsGM Mesangial Cell Growth Me
Supplier:  Lonza Walkersville CC4146
Encompass
MsGM SingleQuot Kit are components sold separately and does not include the basal medium These are part of the MsGM Mesangial Cell Growth Medium BulletKit (Catalog Number CC-3146) The full MsGM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3146 MsGM Mesangial Cell Growth Medium has been specially formulated for the maintenance and expansion of renal mesangial cells MsGM Medium contains 5% serum
Catalog No. 50-004-96373