Learn More
Aviva Systems Biology Corp MSH4 Antibody - middle region (ARP41354_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41354_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MSH4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MSH4. Alternate names: RED2, ADAR3, MSH4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57817