Learn More
Genetex Inc MSH6 Monoclonal Antibody [MSVA-906R] HistoMAX™ (GTX04416) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04416
Encompass_Preferred
The MSH6 antibody [MSVA-906R] HistoMAX™ (GTX04416) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human colorectal adenocarcinoma (COAD) tissue using GTX04416 MSH6 antibody [MSVA-906R] HistoMAX. Colorectal adenocarcinoma showing strong MSH6 positivity in cancer cells and moderate staining in stromal cells.
Catalog No. NC3666878