Learn More
Medchemexpress LLC HY-120079 5mg , MSN-125 CAS:1592908-16-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1200795MG
Encompass_Preferred
HY-120079 5mg MSN-125 CAS: 1592908-16-1 MSN-125 is a potent Bax and Bak oligomerization inhibitor. MSN-125 prevents mitochondrial outer membrane permeabilization (MOMP) with an IC50 of 4 μM. MSN-125 potently inhibits Bax/Bak-mediated apoptosis in HCT-116, BMK Cells, and primary cortical neurons, protects primary neurons against glutamate excitotoxicity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9969