Learn More
TARGETMOL CHEMICALS INC MSP3 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T281185MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MSP3 is an agonist of TRPV1 (EC50 = 0.87 uM) and shows antinociceptive and neuroprotective effects. purity: 99%
Catalog No. 50-258-0548