Learn More
Medchemexpress LLC Msr-Ratio (Msr-green) | 2290635-22-0 | 99.1% | 310.37 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYD12575MG
Encompass_Preferred
Msr-Ratio (Msr-green) is a ratiometric fluorescent probe for methionine sulfoxide reductase. It is utilized to monitor enzyme activity both in vitro and in live cells. The probe converts to its corresponding sulfide form by methionine sulfoxide reductase and exhibits favorable characteristics.
  • Nearly 400-fold fluorescence change
  • Fast response rate (under 30 minutes)
  • Large Stokes shift (120 nm)
  • Green emission (550 nm)
  • Suitable for in vitro and live cell applications
Catalog No. 50-004-28171