Learn More
Medchemexpress LLC MST1/STK4 protein, human | 97.0% | 10 UG

Supplier: Medchemexpress LLC HYP7474210UG
Recombinant human MST1/STK4 protein expressed in Sf9 insect cells and supplied with an N-terminal His tag. Supplied as 10 μg of purified protein for use in biochemical assays and signaling studies; applications include Hippo pathway and apoptosis research. Purity is documented by the certificate of analysis.
- Recombinant human serine/threonine kinase for in vitro studies.
- N-terminal His tag enables affinity purification and detection.
- Produced in Sf9 insect cells to support proper folding.
- Purity greater than 90% as determined by reducing SDS-PAGE.
- Supplied as a small-volume aliquot suitable for biochemical assays.
- Applicable to Hippo pathway and apoptosis-related research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.