Learn More
Medchemexpress LLC Mst1/stk4 protein, human (sf9, his) | >90.0% | 50 UG
SDP
Supplier:  Medchemexpress LLC HYP7474250UG
Encompass_Preferred
Recombinant human MST1/STK4 (serine/threonine-protein kinase 4) produced in Sf9 insect cells with an N-terminal 10xHis tag. Supplied as purified protein for biochemical and cell signaling research; available in small research sizes with supporting COA and SDS documentation.
  • Human-derived serine/threonine kinase 4, amino acids E2-F487.
  • Expressed in Sf9 insect cells.
  • N-terminal 10xHis purification tag.
  • Purity >90.0% as determined by reducing SDS-PAGE.
  • Available in small research sizes (10 μg, 50 μg, 100 μg).
  • Certificate of analysis and safety data sheet available.
Catalog No. 50-002-94315