Learn More
Selleck Chemical LLC MSX-122
SDP
Supplier:  Selleck Chemical LLC S66175MG
Encompass_Preferred
MSX-122 (Q-122) is a novel small molecule and partial CXCR4 antagonist (IC50 10 nM)
Catalog No. 50-000-02812