Learn More
Cayman Chemical MT-DADMe-ImA 25mg
SDP
Supplier:  Cayman Chemical 2971125 MG
Encompass_Preferred
An MTAP inhibitor; increases MTA levels in FaDu human squamous cell carcinoma cells at 1 µM; reduces tumor growth in a FaDu mouse xenograft model at 5, 9, and 21 mg/kg per day for 28 days
Catalog No. 50-298-8899