Learn More
Genetex Inc MTAP Monoclonal Antibody [MSVA-741R] HistoMAX™ (GTX04964) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04964
Encompass_Preferred
The MTAP antibody [MSVA-741R] HistoMAX™ (GTX04964) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human ovarian stroma tissue using GTX04964 MTAP antibody [MSVA-741R] HistoMAX. Moderate predominantly cytoplasmic MTAP staining of ovarian stroma cells.
Catalog No. NC3668421