Learn More
BIOHIPPO mTfrc-sh1-GFP-BSD
SDP
Supplier:  BIOHIPPO BHV194001505
Encompass
mTfrc-sh1-GFP-BSD shRNA lentiviral vector expressing GFP with Blasticidin selection for studies in Mouse Each vial provides a nominal titer of 5x10E6 transducing units TU Validated mouse Tfrc shRNA lentivirus High-titer shRNA lentiviral particles specific for mouse transferrin receptor (Tfrc CD71) The shRNA has been validated to meet or exceed 70% Tfrc knockdown efficiency using a specific fluorescence-based method that is more rapid and reliable than qPCR The shRNA lentivirus is ultra-purified and concentrated to high-titer by PEG precipitation and sucrose gradient centrifugation and ideal for transducing difficult-to-transfect cells including thawed and/or primary cells
Catalog No. 50-003-31360