Learn More
Cayman Chemical mthyl lnolnyl Fluorophos 10mg
SDP
Supplier:  Cayman Chemical 7066410 MG
Encompass_Preferred
An analog of MAFP, which has been widely studied as an inhibitor of phospholipases, FAAH, and as a CB receptor ligand; pharmacology of the γ-linolenyl analog of MAFP has not been completely investigated.
Catalog No. 50-289-3000