Learn More
MSC Mitutoyo Mount, A-6, Nickel-Plated Cast Iron, 9.5 mm, 1 ea
Supplier:  MSC 55314041
Encompass
This mounting fixture provides a reliable solution for securing micrometer heads and linear gages, eliminating the need for laborious and costly custom bracket fabrication. Designed for diverse applications, the fixture is constructed from durable nickel-plated cast iron.
  • Constructed from nickel-plated cast iron
  • Compatible with plain stem units
  • Features a split-clamp design
  • Designed for 9.5 mm stem diameter
Catalog No. 50-005-14781