Learn More
MSC Mitutoyo Radius Gage, Decimal, 0.140 in., 1 ea.
Supplier: MSC 01488568
This radius gage is designed for checking or laying out concave and convex radii in machining, layout, inspection, and patternmaking work. It features a precise 0.140 in. dimension and is finished with a satin or dull-chrome coating for durability and visibility.
- Features five measuring locations for increased versatility
- Includes precisely finished, smooth, and accurate edges
- Identified with clear radius dimension markings
- Designed for easier, faster, and more accurate verification of radius or fillet dimensions